Assay Details
| Gene Symbol: LOC114116198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972725.1 | XP_027828526.1 | 4 - 5 | 658 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114116198| Gene Symbol: LOC114116198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972725.1 | XP_027828526.1 | 4 - 5 | 658 | 82 | |
Set Membership: |
|