Assay Details
| Gene Symbol: NACC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966196.1 | XP_027821997.1 | 5 - 6 | 1427 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NACC2| Gene Symbol: NACC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966196.1 | XP_027821997.1 | 5 - 6 | 1427 | 68 | |
Set Membership: |
|