Assay Details
Gene Symbol: GET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003350.4 | XP_004003399.3 | 4 - 5 | 511 | 83 | |
XM_012103808.3 | XP_011959198.2 | 4 - 5 | 483 | 83 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003350.4 | XP_004003399.3 | 4 - 5 | 511 | 83 | |
XM_012103808.3 | XP_011959198.2 | 4 - 5 | 483 | 83 |