Assay Details
| Gene Symbol: LOC101115398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015867.2 | XP_004015916.2 | 1 - 2 | 380 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115398| Gene Symbol: LOC101115398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015867.2 | XP_004015916.2 | 1 - 2 | 380 | 84 | |
Set Membership: |
|