Assay Details
| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018939.4 | XP_004018988.2 | 2 - 3 | 531 | 77 | |
| XM_015102788.2 | XP_014958274.2 | 2 - 3 | 531 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAPCD1| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018939.4 | XP_004018988.2 | 2 - 3 | 531 | 77 | |
| XM_015102788.2 | XP_014958274.2 | 2 - 3 | 531 | 77 | ||