Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019102.4 | XP_004019151.2 | 2 - 3 | 480 | 98 | |
| Gene Symbol: LOC114109597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958615.1 | XP_027814416.1 | 2 - 3 | 451 | 98 | |
More Information
Set Membership: |
|

ACOT13
Genomic Map