Assay Details
| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020264.4 | XP_004020313.2 | 1 - 2 | 4103 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSYN2A| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020264.4 | XP_004020313.2 | 1 - 2 | 4103 | 84 | |