Assay Details
| Gene Symbol: POLI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020550.4 | XP_004020599.2 | 4 - 5 | 548 | 71 | |
| XM_012103721.3 | XP_011959111.2 | 3 - 4 | 345 | 71 | ||
| XM_012103723.2 | XP_011959113.2 | 3 - 4 | 445 | 71 | ||
| XM_027961056.1 | XP_027816857.1 | 4 - 5 | 452 | 71 | ||
| XM_027961058.1 | XP_027816859.1 | 4 - 5 | 547 | 71 | ||
| XM_027961059.1 | XP_027816860.1 | 4 - 5 | 547 | 71 | ||
More Information
Set Membership: |
|

POLI
Genomic Map