Assay Details
| Gene Symbol: GOLGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021795.4 | XP_004021844.1 | 4 - 5 | 442 | 74 | |
| XM_012106001.3 | XP_011961391.1 | 4 - 5 | 415 | 74 | ||
More Information
Set Membership: |
|

GOLGA7
Genomic Map