Assay Details
| Gene Symbol: LOC101114746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023458.4 | XP_004023507.2 | 9 - 10 | 1557 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114746| Gene Symbol: LOC101114746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023458.4 | XP_004023507.2 | 9 - 10 | 1557 | 89 | |
Set Membership: |
|