Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958365.1 | XP_027814166.1 | 9 - 10 | 2076 | 55 | |
| XM_027958367.1 | XP_027814168.1 | 9 - 10 | 2076 | 55 | ||
More Information
Set Membership: |
|

TEAD3
Genomic Map