Assay Details
| Gene Symbol: CCNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957382.1 | XP_027813183.1 | 7 - 8 | 1052 | 73 | |
| XM_027957383.1 | XP_027813184.1 | 7 - 8 | 1050 | 73 | ||
More Information
Set Membership: |
|

CCNK
Genomic Map