Assay Details
| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181884.3 | XP_012037274.1 | 6 - 7 | 1418 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RDH11| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181884.3 | XP_012037274.1 | 6 - 7 | 1418 | 73 | |
Set Membership: |
|