Assay Details
| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103796.2 | XP_014959282.2 | 8 - 9 | 1114 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MALT1| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103796.2 | XP_014959282.2 | 8 - 9 | 1114 | 63 | |
Set Membership: |
|