Assay Details
| Gene Symbol: STX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972523.1 | XP_027828324.1 | 3 | 675 | 53 | |
| XM_027972524.1 | XP_027828325.1 | 3 | 672 | 53 | ||
| XM_027972525.1 | XP_027828326.1 | 3 | 797 | 53 | ||
| XM_027972526.1 | XP_027828327.1 | 2 | 612 | 53 | ||
More Information
Set Membership: |
|

STX11
Genomic Map