Assay Details
| Gene Symbol: LOC101115551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098173.2 | XP_014953659.2 | 10 - 11 | 1101 | 109 | |
| XM_015098174.2 | XP_014953660.2 | 10 - 11 | 1101 | 109 | ||
| XM_027973646.1 | XP_027829447.1 | 6 - 7 | 610 | 109 | ||
More Information
Set Membership: |
|

LOC101115551
Genomic Map