Assay Details
Gene Symbol: LOC101122608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973511.1 | XP_027829312.1 | 11 - 12 | 927 | 107 | |
GenBank mRNA | FJ958364.1 | - | 11 - 12 | 726 | 107 |

More Information
Set Membership: |
![]() ![]() |
