Assay Details
| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 6 - 7 | 584 | 91 | |
| GenBank mRNA | FJ958364.1 | - | 6 - 7 | 383 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122608| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 6 - 7 | 584 | 91 | |
| GenBank mRNA | FJ958364.1 | - | 6 - 7 | 383 | 91 | |