Assay Details
| Gene Symbol: LOC101120043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961465.1 | XP_027817266.1 | 2 - 3 | 459 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120043| Gene Symbol: LOC101120043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961465.1 | XP_027817266.1 | 2 - 3 | 459 | 95 | |