Assay Details
| Gene Symbol: MLKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189543.2 | XP_012044933.2 | 3 - 4 | 575 | 70 | |
| XM_027977620.1 | XP_027833421.1 | 4 - 5 | 1230 | 70 | ||
| XM_027977621.1 | XP_027833422.1 | 4 - 5 | 737 | 70 | ||
| XM_027977622.1 | XP_027833423.1 | 4 - 5 | 1029 | 70 | ||
More Information
Set Membership: |
|

MLKL
Genomic Map