Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971542.1 | XP_027827343.1 | 1 - 2 | 249 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LARP6| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971542.1 | XP_027827343.1 | 1 - 2 | 249 | 109 | |
Set Membership: |
|