Assay Details
| Gene Symbol: LOC101114079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961988.1 | XP_027817789.1 | 6 - 7 | 813 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114079| Gene Symbol: LOC101114079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961988.1 | XP_027817789.1 | 6 - 7 | 813 | 151 | |
Set Membership: |
|