Assay Details
| Gene Symbol: ZNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099969.2 | XP_014955455.2 | 3 - 4 | 1588 | 88 | |
| XM_027978219.1 | XP_027834020.1 | 4 - 5 | 1615 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF181| Gene Symbol: ZNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099969.2 | XP_014955455.2 | 3 - 4 | 1588 | 88 | |
| XM_027978219.1 | XP_027834020.1 | 4 - 5 | 1615 | 88 | ||