Assay Details
| Gene Symbol: LOC101115853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962283.1 | XP_027818084.1 | 1 - 2 | 74 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027962283.1 transcripts.
Set Membership: |
|

LOC101115853
Genomic Map