Assay Details
| Gene Symbol: GEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966532.1 | XP_027822333.1 | 6 - 7 | 1693 | 63 | |
| XM_027966533.1 | XP_027822334.1 | 7 - 8 | 1360 | 63 | ||
More Information
Set Membership: |
|

GEN1
Genomic Map