Assay Details
| Gene Symbol: LOC101114145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958808.1 | XP_027814609.1 | 2 - 3 | 210 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114145| Gene Symbol: LOC101114145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958808.1 | XP_027814609.1 | 2 - 3 | 210 | 103 | |
Set Membership: |
|