Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189745.3 | XP_012045135.3 | 3 - 4 | 827 | 94 | |
| XM_027977819.1 | XP_027833620.1 | 3 - 4 | 827 | 94 | ||
More Information
Set Membership: |
|

TENT4B
Genomic Map