Assay Details
| Gene Symbol: MCF2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957913.1 | XP_027813714.1 | 30 - 31 | 3717 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCF2L2| Gene Symbol: MCF2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957913.1 | XP_027813714.1 | 30 - 31 | 3717 | 61 | |
Set Membership: |
|