Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002858.3 | XP_004002907.2 | 4 - 5 | 788 | 73 | |
| XM_012188318.2 | XP_012043708.2 | 4 - 5 | 806 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIOX2| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002858.3 | XP_004002907.2 | 4 - 5 | 788 | 73 | |
| XM_012188318.2 | XP_012043708.2 | 4 - 5 | 806 | 73 | ||