Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002858.3 | XP_004002907.2 | 6 - 7 | 1000 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIOX2| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002858.3 | XP_004002907.2 | 6 - 7 | 1000 | 82 | |