Assay Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004078.4 | XP_004004127.1 | 7 - 8 | 1026 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPP| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004078.4 | XP_004004127.1 | 7 - 8 | 1026 | 77 | |
Set Membership: |
|