Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004231.4 | XP_004004280.1 | 4 - 5 | 1092 | 88 | |
| XM_012120584.2 | XP_011975974.1 | 4 - 5 | 1127 | 88 | ||
| XM_012120593.3 | XP_011975983.1 | 2 - 3 | 759 | 88 | ||
More Information
Set Membership: |
|

TRMO
Genomic Map