Assay Details
| Gene Symbol: MRPL55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008395.3 | XP_004008444.1 | 3 - 4 | 522 | 92 | |
| XM_004008396.4 | XP_004008445.2 | 2 - 3 | 741 | 92 | ||
| XM_012177642.3 | XP_012033032.1 | 3 - 4 | 464 | 92 | ||
| XM_027969323.1 | XP_027825124.1 | 3 - 4 | 831 | 92 | ||
| XM_027969324.1 | XP_027825125.1 | 3 - 4 | 467 | 92 | ||
| XM_027969325.1 | XP_027825126.1 | 3 - 4 | 473 | 92 | ||
| XM_027969326.1 | XP_027825127.1 | 3 - 4 | 828 | 92 | ||
More Information
Set Membership: |
|

MRPL55
Genomic Map