Assay Details
| Gene Symbol: LOC101102485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970981.1 | XP_027826782.1 | 6 - 7 | 465 | 69 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101102485| Gene Symbol: LOC101102485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970981.1 | XP_027826782.1 | 6 - 7 | 465 | 69 | |