Assay Details
| Gene Symbol: LOC101103154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009901.4 | XP_004009950.2 | 4 - 5 | 789 | 77 | |
| XM_027971062.1 | XP_027826863.1 | 4 - 5 | 784 | 77 | ||
| XM_027971063.1 | XP_027826864.1 | 4 - 5 | 784 | 77 | ||
More Information
Set Membership: |
|

LOC101103154
Genomic Map