Assay Details
| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010262.4 | XP_004010311.1 | 4 - 5 | 562 | 88 | |
| XM_012180830.3 | XP_012036220.1 | 3 - 4 | 418 | 88 | ||
| XM_012180831.3 | XP_012036221.1 | 3 - 4 | 1392 | 88 | ||
| GenBank mRNA | DQ239677.1 | - | 2 - 3 | 328 | 88 | |
More Information
Set Membership: |
|

CALML4
Genomic Map