Assay Details
| Gene Symbol: HTR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012066.4 | XP_004012115.1 | 3 - 4 | 1051 | 103 | |
| GenBank mRNA | AF039389.1 | - | 1 - 2 | 100 | 103 | |
More Information
Set Membership: |
|

HTR2A
Genomic Map