Assay Details
| Gene Symbol: PRTFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014225.4 | XP_004014274.3 | 4 - 5 | 571 | 68 | |
| XM_012188367.3 | XP_012043757.2 | 4 - 5 | 571 | 68 | ||
| XM_012188371.3 | XP_012043761.2 | 4 - 5 | 571 | 68 | ||
| XM_027976629.1 | XP_027832430.1 | 4 - 5 | 462 | 68 | ||
| XM_027976630.1 | XP_027832431.1 | 4 - 5 | 516 | 68 | ||
More Information
Set Membership: |
|

PRTFDC1
Genomic Map