Assay Details
| Gene Symbol: C15H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979169.1 | XP_027834970.1 | 1 - 2 | 89 | 93 | |
| XM_027979170.1 | XP_027834971.1 | 1 - 2 | 89 | 93 | ||
More Information
Set Membership: |
|

C15H11orf65
Genomic Map