Assay Details
| Gene Symbol: USP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017438.4 | XP_004017487.1 | 4 - 5 | 541 | 67 | |
| XM_012097945.3 | XP_011953335.1 | 4 - 5 | 541 | 67 | ||
More Information
Set Membership: |
|

USP30
Genomic Map