Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103783.3 | XP_011959173.2 | 5 - 6 | 542 | 84 | |
| XM_027960715.1 | XP_027816516.1 | 5 - 6 | 540 | 84 | ||
More Information
Set Membership: |
|

NARS1
Genomic Map