Assay Details
| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187301.3 | XP_012042691.2 | 5 - 6 | 605 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF1B| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187301.3 | XP_012042691.2 | 5 - 6 | 605 | 61 | |
Set Membership: |
|