Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971303.1 | XP_027827104.1 | 18 - 19 | 4053 | 98 | |
XM_027971304.1 | XP_027827105.1 | 17 - 18 | 3667 | 98 | ||
XM_027971305.1 | XP_027827106.1 | 18 - 19 | 3333 | 98 | ||
XM_027971306.1 | XP_027827107.1 | 18 - 19 | 4070 | 98 | ||
XM_027971307.1 | XP_027827108.1 | 17 - 18 | 3647 | 98 | ||
XM_027971308.1 | XP_027827109.1 | 17 - 18 | 3321 | 98 |
