Assay Details
| Gene Symbol: RBM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012132816.3 | XP_011988206.2 | 6 - 7 | 1103 | 71 | |
| XM_012132826.3 | XP_011988216.2 | 6 - 7 | 1101 | 71 | ||
| XM_027964801.1 | XP_027820602.1 | 6 - 7 | 1104 | 71 | ||
| XM_027964802.1 | XP_027820603.1 | 6 - 7 | 1104 | 71 | ||
| XM_027964803.1 | XP_027820604.1 | 6 - 7 | 1104 | 71 | ||
| XM_027964804.1 | XP_027820605.1 | 6 - 7 | 1098 | 71 | ||
More Information
Set Membership: |
|

RBM45
Genomic Map