Assay Details
| Gene Symbol: SNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095947.2 | XP_014951433.1 | 1 - 2 | 143 | 62 | |
| XM_015095948.2 | XP_014951434.1 | 1 - 2 | 140 | 62 | ||
More Information
Set Membership: |
|

SNX2
Genomic Map