Assay Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100072.2 | XP_014955558.2 | 4 - 5 | 548 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF577| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100072.2 | XP_014955558.2 | 4 - 5 | 548 | 87 | |