Assay Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972344.1 | XP_027828145.1 | 8 - 9 | 1156 | 77 | |
| XM_027972348.1 | XP_027828149.1 | 8 - 9 | 1067 | 77 | ||
More Information
Set Membership: |
|

LRRC39
Genomic Map