Assay Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002561.4 | XP_004002610.2 | 3 - 4 | 820 | 92 | |
| XM_012182536.3 | XP_012037926.2 | 3 - 4 | 822 | 92 | ||
| XM_027975799.1 | XP_027831600.1 | 2 - 3 | 237 | 92 | ||
More Information
Set Membership: |
|

PMVK
Genomic Map