Assay Details
| Gene Symbol: HSD17B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964729.1 | XP_027820530.1 | 7 - 8 | 470 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSD17B3| Gene Symbol: HSD17B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964729.1 | XP_027820530.1 | 7 - 8 | 470 | 68 | |
Set Membership: |
|