Assay Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007795.4 | XP_004007844.2 | 6 - 7 | 808 | 70 | |
| XM_012176520.3 | XP_012031910.2 | 6 - 7 | 808 | 70 | ||
More Information
Set Membership: |
|

DBF4
Genomic Map